1-Naphthyl phosphate monosodium salt monohydrate 98+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1-Naphthylphosphoric Acid Monosodium Salt Monohydrate, Monosodium 1-Naphthyl Phosphate Monohydrate, Sodium 1-Naphthyl Hydrogen Phosphate Monohydrate, Phosphoric Acid 1-Naphthyl Ester Monosodium Salt Monohydrate

226890050 226890010 226890250 226891000
ACRO226890050EA 91.4 EUR
ACRO226890050 ACRO226890010 ACRO226890250 ACRO226891000
1-Naphthyl phosphate monosodium salt monohydrate 98+%
1-Naphthyl phosphate monosodium salt monohydrate
Formule: C₁₀H₈NaO₄P·1H₂O
MW: 264.15 g/mol
Smeltpunt: 189…191 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00150615
CAS nummer: 81012-89-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless to light yellow
Titration with HClO4 ≥98.0 %
Impurity ≤100 ppm Free 1-Naphthol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR