X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 99%, pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

227770100 227771000 227770025 227775000
ACRO227770100EA 19.9 EUR
ACRO227770100 ACRO227771000 ACRO227770025 ACRO227775000
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 99%, pure
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Formule: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
Smeltpunt: 230 °C (ontbinding)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005666
CAS nummer: 7240-90-6
EINECS: 230-640-8

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
HPLC ≥98.5 %
Specific optical rotation -60° to -64° (20°C, 589 nm) (c=1, H2O/DMF 1/1)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR