Chloramine-T (sodium salt) trihydrate 97+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt

227850010 227850250 227852500 227850050
ACRO227850010EA 105 EUR
ACRO227850010 ACRO227850250 ACRO227852500 ACRO227850050
Chloramine-T (sodium salt) trihydrate 97+%
Chloramine-T (sodium salt) trihydrate
Formule: CH₃C₆H₄SO₂NClNa·3H₂O
Dichtheid: 1,43 g/cm³ (20 °C)
Vlampunt: 192 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149066
CAS nummer: 7080-50-4
EINECS: 204-854-7
UN: 3263
ADR: 8,III
Merck Index: 13,02085

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Titration Iodometric 97.0 to 103.0 %
Appearance of solution (5% in water) Clear solution
pH 9.0 to 11.0
Color of solution ≤25 APHA (5 % in water)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR