TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 99+%, extra pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl

228030010 228031000 228032500 228030051
ACRO228030010EA 296 EUR
ACRO228030010 ACRO228031000 ACRO228032500 ACRO228030051
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 99+%, extra pure
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)
Formule: NH₂C(CH₂OH)₃·HCl
Kookpunt: 225 °C
Smeltpunt: 150…151 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012590
CAS nummer: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 150 to 153 °C
Sulfated ash ≤0.1 %
Solubility (1 % in water) Clear colorless
pH 3.5 to 5.0 (1 % at 25 °C)
Titration with HClO4 ≥99.0 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR