Pyridoxal-5'-phosphate monohydrate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

228170010 228170050 228170250
ACRO228170010EA 39.4 EUR
ACRO228170010 ACRO228170050 ACRO228170250
Pyridoxal-5'-phosphate monohydrate 99%
Pyridoxal-5'-phosphate monohydrate
Formule: C₈H₁₀NO₆P·1H₂O
Smeltpunt: 140…143 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00149414
CAS nummer: 41468-25-1

Order Now

Specification Test Results

Appearance Off-white to yellow Powder
Infrared spectrum Conforms
Water ≤10 %
HPLC ≥98.5 %
Phosphorus (P) ≤0.2 % (P inorganic)
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR