5-Bromo-2'-deoxyuridine (broxuridine) 99+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

228590010 228590025 228595000 228590100
ACRO228590010EA 87.4 EUR
ACRO228590010 ACRO228590025 ACRO228595000 ACRO228590100
5-Bromo-2'-deoxyuridine (broxuridine) 99+%
5-Bromo-2'-deoxyuridine (broxuridine)
Formule: C₉H₁₁BrN₂O₅
Smeltpunt: 191…194 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006529
CAS nummer: 59-14-3
EINECS: 200-415-9

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤2 % (K.F.)
HPLC ≥99.0 %
Ratio of Absorbances 1.45 to 1.55 (A290/A260)
Microbial contamination ≤100 CFU/g (Total count)
Bacterial endotoxins ≤0.1 EU/mg (LAL-test)
Related substances ≤1.0 % (HPLC) Total impurities
Specific optical rotation +20° to +24° (20°C, 589 nm) (c=1, H2O)
Heavy metals (as Pb) ≤10 ppm
Residual solvents ≤0.1 % (GC) Methanol

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR