Creatinine 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

228940250 228940500 228942500
ACRO228940250EA 61.3 EUR
ACRO228940250 ACRO228940500 ACRO228942500
Creatinine 98%
Creatinine
Formule: C₄H₇N₃O
Vlampunt: 290 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00059730
CAS nummer: 60-27-5
EINECS: 200-466-7

Order Now

Specification Test Results

Appearance White to yellowish-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105°C)
Sulfated ash ≤0.2 %
Solubility (5 % in water) Clear colorless to slightly yellow
Titration with HClO4 97.5 to 102.5 % (on dried substance)
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR