L-Valine methyl ester hydrochloride 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

229260100 229260500
ACRO229260100EA 53.8 EUR
ACRO229260100 ACRO229260500
L-Valine methyl ester hydrochloride 99%
L-Valine methyl ester hydrochloride

Appearance: White Powder or crystals

Formule: C₆H₁₄ClNO₂
Smeltpunt: 171…173 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012497
CAS nummer: 6306-52-1

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Solubility (5 % in water) Clear colorless to light yellow
Specific optical rotation +14° to +17° (20°C, 589 nm) (c=2, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR