3,3',5,5'-Tetramethylbenzidine (TMB) 99+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

229280050 229280010 229280250
ACRO229280050EA 257 EUR
ACRO229280050 ACRO229280010 ACRO229280250
3,3',5,5'-Tetramethylbenzidine (TMB) 99+%
3,3',5,5'-Tetramethylbenzidine (TMB)
Formule: C₁₆H₂₀N₂
Kookpunt: 402 °C (1013 hPa)
Smeltpunt: 167…170 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007748
CAS nummer: 54827-17-7
EINECS: 259-364-6

Order Now

Specification Test Results

Appearance White to cream to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 167 to 172 °C
Loss on drying ≤0.5 %
Sulfated ash ≤0.2 %
HPLC ≥99.0 %
Titration with HClO4 98.0 to 102.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR