Eosin B, high purity biological stain

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91

229650250 229651000
ACRO229650250EA 33.8 EUR
ACRO229650250 ACRO229651000
Eosin B, high purity biological stain
Eosin B
Formule: C₂₀H₆Br₂N₂Na₂O₉
MW: 624.06 g/mol
Smeltpunt: >310 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005041
CAS nummer: 548-24-3
EINECS: 208-943-1
Merck Index: 12,03638

Order Now

Specification Test Results

Appearance Dark brown Crystalline powder
Infrared spectrum Conforms
UV Lambda max 511 to 520 nm (in water + 1% Na2CO3)
Absorption Ratio P-15/P+15 0.96 to 1.22


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR