Magnesium stearate (3,8 - 5,0% Mg)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Stearic acid Magnesium(II) salt, Magnesium(II) Stearate, Stearic acid magnesium salt

240760010 240760250 240760050
ACRO240760010EA 44.4 EUR
ACRO240760010 ACRO240760250 ACRO240760050
Magnesium stearate (3,8 - 5,0% Mg)
Magnesium stearate
Formule: C₃₆H₇₀MgO₄
Smeltpunt: 130…140 °C
Dichtheid: 1,03 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036391
CAS nummer: 557-04-0
EINECS: 209-150-3
Merck Index: 13,05714

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Titration Complexometric 3.8 to 5.0 % (Mg) On dried substance
Loss on drying ≤6 % (1 g, 105°C)
GC ≥90.0 % rel. content of stearic and palmitic acid
Chloride (Cl) ≤1000 ppm
Sulfate (SO4) ≤1 %
Cadmium (Cd) ≤3 ppm
Lead (Pb) ≤10 ppm
Nickel (Ni) ≤5 ppm
Heavy metals (as Pb) ≤40 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR