Methyl trifluoroacetate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Trifluoroacetic methyl ester

243410250 243411000
ACRO243410250EA 76.9 EUR
ACRO243410250 ACRO243411000
Methyl trifluoroacetate 99%
Methyl trifluoroacetate
Formule: C₃H₃F₃O₂
MW: 128.05 g/mol
Kookpunt: 43…43.5 °C (760 mmHg)
Smeltpunt: –78 °C
Dichtheid: 1.273 g/cm³
Vlampunt: –7 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000417
CAS nummer: 431-47-0
EINECS: 207-074-5
UN: 2924
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR