2-Hydroxypyridine-N-oxide 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

243790250 243791000 243795000
ACRO243790250EA 67 EUR
ACRO243790250 ACRO243791000 ACRO243795000
2-Hydroxypyridine-N-oxide 97%
2-Hydroxypyridine-N-oxide
Formule: C₅H₅NO₂
Smeltpunt: 147…152 °C
Vlampunt: 100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006195
CAS nummer: 13161-30-3

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 147 to 152 °C
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR