5-Fluorocytosine 99+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

258340010 258340050 258342500
ACRO258340010EA 169 EUR
ACRO258340010 ACRO258340050 ACRO258342500
5-Fluorocytosine 99+%
5-Fluorocytosine
Formule: C₄H₄FN₃O
Kookpunt: 298 °C (1013 hPa)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006035
CAS nummer: 2022-85-7
EINECS: 217-968-7
Merck Index: 13,04151

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1.0 % (1 g, 105°C)
Titration with HClO4 ≥99.0 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR