Triphosgene 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

258950050 258950100 258950500 258952500
ACRO258950050EA 41.5 EUR
ACRO258950050 ACRO258950100 ACRO258950500 ACRO258952500
Triphosgene 99%
Triphosgene
Formule: C₃Cl₆O₃
Kookpunt: 203…206 °C (760 mmHg)
Smeltpunt: 77…81 °C
Dichtheid: 1,78 g/cm³
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00062848
CAS nummer: 32315-10-9
EINECS: 250-986-3
UN: 2928
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 77 to 81 °C
Assay ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR