Butylbutyraat 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Butyric acid butyl ester

259590010 259590050 259591000
ACRO259590010EA 83.1 EUR
ACRO259590010 ACRO259590050 ACRO259591000
Butylbutyraat 98%
Butylbutyraat
Formule: C₈H₁₆O₂
MW: 144.21 g/mol
Kookpunt: 166
Smeltpunt: –92 °C
Dichtheid: 0.8675 g/cm³
Vlampunt: 49 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009450
CAS nummer: 109-21-7
EINECS: 203-656-8
UN: 1993
ADR: 3 III
Merck Index: 13,01555

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4070 (20°C, 589 nm)
GC ≥97.5 %
Specific gravity (25°C) 0.866 to 0.869
Acid value ≤1 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR