2-Phenyl-5-benzimidazolesulphonic acid 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Ensulizole, Phenylbenzimidazole sulphonic acid, 1H-Benzimidazole-5-sulphonic acid

259700250 259700050 259702500
ACRO259700250EA 40.4 EUR
ACRO259700250 ACRO259700050 ACRO259702500
2-Phenyl-5-benzimidazolesulphonic acid 98%
2-Phenyl-5-benzimidazolesulphonic acid

CAS No.: 27503-81-7

Formule: C₁₃H₁₀N₂O₃S
MW: 274.29 g/mol
Smeltpunt: >300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00053007
CAS nummer: 27503-81-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤7 %
Titration excess NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR