3,4-Dibromothiophene 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

264520010 264520050 264520250
ACRO264520010EA 36.4 EUR
ACRO264520010 ACRO264520050 ACRO264520250
3,4-Dibromothiophene 97%
3,4-Dibromothiophene
Formule: C₄H₂Br₂S
Kookpunt: 221…222 °C (760 mmHg)
Smeltpunt: 4…5 °C
Dichtheid: 2,13 g/cm³
Vlampunt: >110 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005465
CAS nummer: 3141-26-2
EINECS: 221-546-8
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.6380 to 1.6420 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR