2,2,4-Trimethylpentane 99.5% HPLC

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

268810010 268810025
ACRO268810010EA 78.2 EUR
ACRO268810010 ACRO268810025
2,2,4-Trimethylpentane 99.5% HPLC
2,2,4-Trimethylpentane
Formule: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Kookpunt: 99.2
Smeltpunt: –107 °C
Dichtheid: 0.69 g/cm³
Vlampunt: –12 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008943
CAS nummer: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3 II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.01 %
GC ≥99.4 %
UV 1 cm cell vs HPLC-grade water
Residue after evaporation ≤0.0002 %
Acidity ≤0.0002 meq/g
Filtered to 0.2 micrometer Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR