Triphenylphosphonium bromide 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Triphenylphosphine hydrobromide

269090250 269091000 269090050
ACRO269090250EA 73.2 EUR
ACRO269090250 ACRO269091000 ACRO269090050
Triphenylphosphonium bromide 99%
Triphenylphosphonium bromide
Formule: C₁₈H₁₆BrP
MW: 343.2 g/mol
Smeltpunt: 196…199 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00035107
CAS nummer: 6399-81-1
EINECS: 229-012-6
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR