Disodium hydrogen phosphate dodecahydrate 98.5% for analysis

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

271060010 271060025 271060100
ACRO271060010EA 45.8 EUR
ACRO271060010 ACRO271060025 ACRO271060100
Disodium hydrogen phosphate dodecahydrate 98.5% for analysis
Disodium hydrogen phosphate dodecahydrate
Formule: Na₂HPO₄·12H₂O
Smeltpunt: 35 °C
Dichtheid: 1,52 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149181
CAS nummer: 10039-32-4
EINECS: 231-448-7
Merck Index: 13,08733

Order Now

Specification Test Results

Appearance White Crystals
pH 9 to 9.4 (5 % in water)
Titration with HCl 98.0 to 102.0 %
Chloride (Cl) ≤5 ppm
Sulfate (SO4) ≤50 ppm
Arsenic (As) ≤0.5 ppm
Cadmium (Cd) ≤5 ppm
Calcium (Ca) ≤10 ppm
Chromium (Cr) ≤5 ppm
Cobalt (Co) ≤5 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤5 ppm
Magnesium (Mg) ≤10 ppm
Manganese (Mn) ≤5 ppm
Nickel (Ni) ≤5 ppm
Potassium (K) ≤100 ppm
Zinc (Zn) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Impurity Mono- or tribasic salts: passes test
Insoluble matter ≤0.005 % (in water)
Nitrogen compounds (as N) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR