Tris(trimethylsilyl)silane 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

291060025 291060250
ACRO291060025EA 62.8 EUR
ACRO291060025 ACRO291060250
Tris(trimethylsilyl)silane 96%
Tris(trimethylsilyl)silane
Formule: C₉H₂₈Si₄
Kookpunt: 82…84 °C (12 mmHg)
Dichtheid: 0,8 g/cm³
Vlampunt: 55 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00077893
CAS nummer: 1873-77-4
UN: 1993
ADR: 3,III
Merck Index: 12,09886

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4890 to 1.4910 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR