Dimethylsulfoxide 99.8+%, extra pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

295520010 295522500 295520025 295520250
ACRO295520010EA 78.9 EUR
ACRO295520010 ACRO295522500 ACRO295520025 ACRO295520250
Dimethylsulfoxide 99.8+%, extra pure
Dimethylsulfoxide
Formule: (CH₃)₂SO
MW: 78.14 g/mol
Kookpunt: 189 °C (760 mmHg)
Smeltpunt: 18.4 °C
Dichtheid: 1.1 g/cm³
Vlampunt: 87 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002089
CAS nummer: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 %
GC ≥99.80 %
Color scale ≤10 APHA
Residue after evaporation ≤0.003 %
Acidity ≤0.04 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR