Rhodamine B 98+%, pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride

296570100 296570250 296570010 296571000
ACRO296570100EA 29.3 EUR
ACRO296570100 ACRO296570250 ACRO296570010 ACRO296571000
Rhodamine B 98+%, pure
Rhodamine B
Formule: C₂₈H₃₁ClN₂O₃
MW: 479.02 g/mol
Smeltpunt: 210…211 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011931
CAS nummer: 81-88-9
EINECS: 201-383-9
Merck Index: 13,08266

Order Now

Specification Test Results

Appearance Dark green Powder or crystals
Infrared spectrum Conforms
HPLC ≥98.0 %
Moisture ≤2.0 % (105°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR