5-Nitrosalicylic acid 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

300480050 300480250 300482500
ACRO300480050EA 21 EUR
ACRO300480050 ACRO300480250 ACRO300482500
5-Nitrosalicylic acid 99%
5-Nitrosalicylic acid
Formule: C₇H₅NO₅
Smeltpunt: 228…231 °C
Dichtheid: 1,65 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007338
CAS nummer: 96-97-9
EINECS: 202-548-8
Merck Index: 12,06730

Order Now

Specification Test Results

Appearance Off-white to yellow to beige to orange to tan Powder
Infrared spectrum Conforms
Melting point 228 to 233 °C
HPLC ≥98.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR