L-Proline methyl ester hydrochloride 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

303000050 303000010 303000250
ACRO303000050EA 80.6 EUR
ACRO303000050 ACRO303000010 ACRO303000250
L-Proline methyl ester hydrochloride 98%
L-Proline methyl ester hydrochloride

CAS No.: 2133-40-6

Formule: C₆H₁₁NO₂·HCl
Smeltpunt: 71 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00012708
CAS nummer: 2133-40-6

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Water ≤0.5 % (K.F.)
Specific optical rotation -31° to -35° (20°C, 589 nm) (c=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR