3,5-Bis(trifluoromethyl)benzylamine 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

305640010 305640050
ACRO305640010EA 37.9 EUR
ACRO305640010 ACRO305640050
3,5-Bis(trifluoromethyl)benzylamine 95%
3,5-Bis(trifluoromethyl)benzylamine
Formule: C₉H₇F₆N
Kookpunt: 82…84 °C (15 mmHg)
Smeltpunt: 46…55 °C
Vlampunt: 78 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009909
CAS nummer: 85068-29-7
EINECS: 285-290-9
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals or mass and/or chunks
Infrared spectrum Conforms
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR