4'-tert-Butylacetophenone 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

308740050 308740010
ACRO308740050EA 73 EUR
ACRO308740050 ACRO308740010
4'-tert-Butylacetophenone 98%
4'-tert-Butylacetophenone
Formule: C₁₂H₁₆O
Kookpunt: 107…108 °C (5 mmHg)
Smeltpunt: 17…18 °C
Dichtheid: 0,96 g/cm³
Vlampunt: 29 °C
MDL Nummer: MFCD00017256
CAS nummer: 943-27-1
EINECS: 213-399-3
UN: 1224

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5220 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR