Silver 99.9% (trace metals basis), draad, Ø 0.25 mm , 1ea = 10.4gr

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

317730010
ACRO317730010EA 115 EUR
ACRO317730010
Silver 99.9% (trace metals basis), draad, Ø 0.25 mm , 1ea = 10.4gr
Silver
Formule: Ag
Kookpunt: 2212 °C (1013 hPa)
Smeltpunt: 961 °C
Dichtheid: 10,5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now

Specification Test Results

Appearance Silver white Wire
Total trace metal impurities ≤0.1 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR