3,5-Bis(trifluoromethyl)phenyl isothiocyanate 99+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

319520010 319520050
ACRO319520010EA 30.6 EUR
ACRO319520010 ACRO319520050
3,5-Bis(trifluoromethyl)phenyl isothiocyanate 99+%
3,5-Bis(trifluoromethyl)phenyl isothiocyanate
Formule: C₉H₃F₆NS
Kookpunt: 63 °C (1,5 mmHg)
Dichtheid: 1,485 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00040838
CAS nummer: 23165-29-9
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear to slightly hazy colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4980 to 1.5020 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR