2,4-Dimethylpyrrole 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

319650050 319650010 319650250
ACRO319650050EA 121 EUR
ACRO319650050 ACRO319650010 ACRO319650250
2,4-Dimethylpyrrole 97%
2,4-Dimethylpyrrole
Formule: C₆H₉N
Kookpunt: 165…167 °C
Dichtheid: 0,924 g/cm³
Vlampunt: >109 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00192088
CAS nummer: 625-82-1
EINECS: 210-912-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4950 to 1.4990 (20°C, 589 nm)
GC ≥96.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR