Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

321270050 321270100
ACRO321270050EA 116 EUR
ACRO321270050 ACRO321270100
Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
Dichloromethane-[D₂]
Formule: CD₂Cl₂
Kookpunt: 40 °C (1013 hPa)
Smeltpunt: –97 °C
Dichtheid: 1,36 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000882
CAS nummer: 1665-00-5
EINECS: 216-776-0
UN: 1593
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.02 % (Coulometric)
D-Enrichment ≥99.75 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR