(Bromomethyl)triphenylphosphonium bromide 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

321480050 321480250
ACRO321480050EA 33.1 EUR
ACRO321480050 ACRO321480250
(Bromomethyl)triphenylphosphonium bromide 98%
(Bromomethyl)triphenylphosphonium bromide
Formule: C₁₉H₁₇Br₂P
Smeltpunt: 230…235 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011864
CAS nummer: 1034-49-7
EINECS: 213-857-2

Order Now

Specification Test Results

Appearance White to light beige to pink Powder or crystals
Infrared spectrum Conforms
Melting point 232 to 242 °C
Titration Argentometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR