Aceton 99.9% (by GC, corrected for water content), ECD tested halocarbons free grade for residue analysis

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

326700010 326700025
ACRO326700010EA 36.6 EUR
ACRO326700010 ACRO326700025
Aceton 99.9% (by GC, corrected for water content), ECD tested halocarbons free grade for residue analysis
Aceton
Formule: CH₃COCH₃
Kookpunt: 56,2 °C (1013 hPa)
Smeltpunt: –95 °C
Dichtheid: 0,79 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008765
CAS nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
Bereik: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.2 % (Coulometric)
GC 99.9 to 100 % (on anhydrous basis)
Color scale ≤10 APHA
GC/ECD (as lindane) ≤2 ng/l (EOX suitab.)
GC/ECD ≤5 ng/l (volatile imp. (as E.Dibromide))
Residue after evaporation ≤0.0002 %
Acidity (CH3COOH) ≤0.002 %

Specificaties

Environmentally Preferable Veiligere gezondheid van mens en milieu
Milieu certificering Chem21 Solvent Selection Guide recommended after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR