n-Hexaan 95% for residue analysis, for trace analysis of polyaromatic hydrocarbons

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

326780010 326780025
ACRO326780010EA 63.8 EUR
ACRO326780010 ACRO326780025
n-Hexaan 95% for residue analysis, for trace analysis of polyaromatic hydrocarbons
n-Hexaan
Formule: H₃C(CH₂)₄CH₃
Kookpunt: 69 °C (760 mmHg)
Smeltpunt: –95 °C
Dichtheid: 0,659 g/cm³
Vlampunt: –22 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009520
CAS nummer: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.005 % (Coulometric)
GC ≥95 %
UV at 270 nm A: ≤0.004
Color scale ≤5 APHA
Residue after evaporation ≤0.0002 %
Acidity (CH3COOH) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR