Ninhydrin for TLC

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate

327022400
ACRO327022400EA 92.1 EUR
ACRO327022400
Ninhydrin for TLC
Ninhydrin

Ozone friendly ready, to use spray for TLC

Formule: C₉H₆O₄
MW: 178.14 g/mol
Kookpunt: 351
Smeltpunt: 250…258 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003791
CAS nummer: 485-47-2
EINECS: 207-618-1
UN: 1950
Merck Index: 13,06582
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ff1a5cd0c772262dac61a6c63dbdeca97baa2c&vid=61d6c7dbd1fe019587f026a878ce0951ce80fcc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Additional info Ozone friendly


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR