Tricine 99+% voor de biochemie

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

327831000 327830250 327830010 327830050
ACRO327831000EA 98 EUR
ACRO327831000 ACRO327830250 ACRO327830010 ACRO327830050
Tricine 99+% voor de biochemie
Tricine
Formule: (HOCH₂)₃CNHCH₂CO₂H
Dichtheid: 1,523 g/cm³ (19.85 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004277
CAS nummer: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
UV at 280 nm A: ≤0.02
pH 4.6 to 5.6 (1 % in water at 25°C)
Titration with HClO4 ≥99.0 %
Lead (Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR