n-Hexaan 97% for electronic use (MOS) residue-free

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

327890010
ACRO327890010EA 64.5 EUR
ACRO327890010
n-Hexaan 97% for electronic use (MOS) residue-free
n-Hexaan
Formule: H₃C(CH₂)₄CH₃
Kookpunt: 69 °C (1013 hPa)
Smeltpunt: –95 °C
Dichtheid: 0,659 g/cm³
Vlampunt: –22 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009520
CAS nummer: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.01 % (Coulometric)
GC ≥96 %
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Acidity ≤0.002 %
Chloride (Cl) ≤2 ppm
Arsenic (As) ≤10 ppb
Barium (Ba) ≤20 ppb
Copper (Cu) ≤10 ppb
Iron (Fe) ≤50 ppb
Heavy metals (as Pb) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR