Sphingosine-1-phosphate 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

328520010
ACRO328520010EA 200 EUR
ACRO328520010
Sphingosine-1-phosphate 96%
Sphingosine-1-phosphate

Appearance: White to off-white to light yellow Powder or solid

Formule: C₁₈H₃₈NO₅P
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00270077
CAS nummer: 26993-30-6

Order Now

Specification Test Results

Appearance White to light yellow to light beige Powder or solid
NMR (1H-NMR) Consistent with structure
Purity ≥95.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR