n-Dodecyl-β-D-maltoside 99%, high purity

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

329370010 329370050 329370250
ACRO329370010EA 191 EUR
ACRO329370010 ACRO329370050 ACRO329370250
n-Dodecyl-β-D-maltoside 99%, high purity
n-Dodecyl-β-D-maltoside
Formule: C₂₄H₄₆O₁₁
Smeltpunt: 217 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00043012
CAS nummer: 69227-93-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥98.5 % (ELSD)
Specific optical rotation +45° to +49.5° (20°C, 589 nm) (c=1, water)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR