Diethylphosphonoacetic acid 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-(Diethoxyphosphoryl)acetic acid, Diethyl (hydroxycarbonylmethyl)phosphonate

330670050 330670250
ACRO330670050EA 31 EUR
ACRO330670050 ACRO330670250
Diethylphosphonoacetic acid 98%
Diethylphosphonoacetic acid
Formule: C₆H₁₃O₅P
MW: 196.139 g/mol
Kookpunt: 150 °C
Dichtheid: 1.22 g/cm³
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00192032
CAS nummer: 3095-95-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR