5-Bromo-2-hydroxybenzyl alcohol 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 4-Bromo-2-(hydroxymethyl)phenol, (5-Bromo-2-hydroxyphenyl)methanol

331480050 331480250
ACRO331480050EA 58.7 EUR
ACRO331480050 ACRO331480250
5-Bromo-2-hydroxybenzyl alcohol 98%
5-Bromo-2-hydroxybenzyl alcohol
Formule: C₇H₇BrO₂
MW: 203.04
Kookpunt: 302
Smeltpunt: 106…111 °C
MDL Nummer: MFCD00004618
CAS nummer: 2316-64-5
EINECS: 219-026-0
Merck Index: 12,01460

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 106 to 111 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR