Polysorbate 40

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

334140010 334140250 334142500 334140025
ACRO334140010EA 0 EUR
ACRO334140010 ACRO334140250 ACRO334142500 ACRO334140025
Polysorbate 40
Polysorbate 40
Formule: C₆₂H₁₂₂O₂₆
Kookpunt: ? 100 °C (1013 hPa)
Smeltpunt: ~ 0,1 °C
Dichtheid: 1,05 g/cm³
Vlampunt: >200 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00165345
CAS nummer: 9005-66-7

Order Now

Specification Test Results

Appearance Clear to hazy light yellow to dark yellow Viscous liquid or paste
Infrared spectrum Conforms
Acid value ≤3 mg KOH/g
Hydroxyl value 89 to 105 mg KOH/g
Saponification value 41 to 55 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR