(+)-Dibenzoyl-L-tartaric anhydride 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

336030050 336030250
ACRO336030050EA 37.5 EUR
ACRO336030050 ACRO336030250
(+)-Dibenzoyl-L-tartaric anhydride 98%
(+)-Dibenzoyl-L-tartaric anhydride

Appearance: White to off-white Crystalline powder

Formule: C₁₈H₁₂O₇
Smeltpunt: 174 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00067102
CAS nummer: 64339-95-3
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 192 to 196 °C
Titration by coupling ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR