D-(-)-Luciferin

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

337050500 337050100
ACRO337050500EA 634 EUR
ACRO337050500 ACRO337050100
D-(-)-Luciferin
D-(-)-Luciferin
Formule: C₁₁H₈N₂O₃S₂
Smeltpunt: 200 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00042929
CAS nummer: 2591-17-5
EINECS: 219-981-3

Order Now

Specification Test Results

Appearance Off-white to light yellow Powder
Infrared spectrum Conforms
Water ≤2.0 % (K.F.)
HPLC ≥97.5 %
UV Lambda max 266 to 270 nm (c=1, CH3OH)
Specific optical rotation -32.0° to -36.0° (20°C, 589 nm) (c=1, DMF)
Molar extinction coefficient ≥6900 l/(mol.cm) (266 to 270 nm, c=1, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR