(R)-(+)-2-Hydroxy-3-phenylpropionic acid 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

344580010 344580050
ACRO344580010EA 48.3 EUR
ACRO344580010 ACRO344580050
(R)-(+)-2-Hydroxy-3-phenylpropionic acid 98%
(R)-(+)-2-Hydroxy-3-phenylpropionic acid
Formule: C₉H₁₀O₃
Smeltpunt: 121…125 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00078062
CAS nummer: 7326-19-4
EINECS: 230-803-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 121 to 125 °C
Specific optical rotation +17° to +20° (20 °C, 589 nm) (c=1, CH3OH)
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR