(Ethylthio)trimethylsilane 90%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

345000010 345000100
ACRO345000010EA 33.7 EUR
ACRO345000010 ACRO345000100
(Ethylthio)trimethylsilane 90%
(Ethylthio)trimethylsilane

Appearance: Clear colorless Liquid

Formule: C₅H₁₄SSi
Kookpunt: 69…70 °C (100 mmHg)
Dichtheid: 0,83 g/cm³
Vlampunt: 16 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042890
CAS nummer: 5573-62-6
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4470 to 1.4510 (20°C, 589 nm)
GC ≥88 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR