2-Acetylcyclohexanone 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

346190050 346190250
ACRO346190050EA 33.1 EUR
ACRO346190050 ACRO346190250
2-Acetylcyclohexanone 97%
2-Acetylcyclohexanone
Formule: C₈H₁₂O₂
Dichtheid: 1,063 g/cm³
Vlampunt: 79 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001633
CAS nummer: 874-23-7
EINECS: 212-858-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5070 to 1.5130 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR