Acriflavine hydrochloride

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

346840100 346840250 346841000
ACRO346840100EA 47.6 EUR
ACRO346840100 ACRO346840250 ACRO346841000
Acriflavine hydrochloride
Acriflavine hydrochloride

Appearance: Orange-red to red-brown Powder

Formule: C₂₇H₂₈Cl₄N₆
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00069039
CAS nummer: 69235-50-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Orange-red to red-brown Powder
Infrared spectrum Conforms
Loss on drying ≤7 % (105°C, 1 h)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR