Ethylene glycol monostearate

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

346951000 346952500
ACRO346951000EA 30.3 EUR
ACRO346951000 ACRO346952500
Ethylene glycol monostearate
Ethylene glycol monostearate

Appearance: White to light yellow Flakes

Formule: C₂₀H₄₀O₃
Kookpunt: 149 °C
Smeltpunt: 55…60 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00051465
CAS nummer: 111-60-4

Order Now

Specification Test Results

Appearance White to light yellow Flakes
Infrared spectrum Conforms
Melting point 55 to 60 °C
Additional info compound is a mixture of various chain-length esters and di(ethylene glycol) ester


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR